Assay Details
| Gene Symbol: LOC489591 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005620396.3 | XP_005620453.1 | 1732 | 76 | ||
| XM_005620398.3 | XP_005620455.1 | 1629 | 76 | |||
| XM_014113819.2 | XP_013969294.1 | 1732 | 76 | |||
| XM_022418909.1 | XP_022274617.1 | 1602 | 76 | |||
| XM_843606.5 | XP_848699.1 | 1738 | 76 | |||
More Information
Set Membership: |
|

LOC489591
Genomic Map