Assay Details
| Gene Symbol: RDH16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005625527.3 | XP_005625584.1 | 924 | 79 | ||
| XM_005625528.3 | XP_005625585.1 | 992 | 79 | |||
| XM_005625529.3 | XP_005625586.1 | 733 | 79 | |||
| XM_005625530.3 | XP_005625587.1 | 795 | 79 | |||
| XM_014117166.2 | XP_013972641.1 | 1513 | 79 | |||
| XM_014117167.2 | XP_013972642.1 | 1146 | 79 | |||
| XM_022424079.1 | XP_022279787.1 | 785 | 79 | |||
More Information
Set Membership: |
|

RDH16
Genomic Map