Assay Details
| Gene Symbol: TSSK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022421841.1 | XP_022277549.1 | 1 - 2 | 567 | 70 | |
| XM_022421842.1 | XP_022277550.1 | 1 - 2 | 566 | 70 | ||
| XM_022421845.1 | XP_022277553.1 | 1 - 2 | 564 | 70 | ||
| XM_022421846.1 | XP_022277554.1 | 1 - 2 | 564 | 70 | ||
| XM_537386.6 | XP_537386.1 | 1 - 2 | 559 | 70 | ||
| XM_844938.5 | XP_850031.1 | 1 - 2 | 559 | 70 | ||
More Information
Set Membership: |
|

TSSK4
Genomic Map