Assay Details
| Gene Symbol: SLC22A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014117710.2 | XP_013973185.1 | 9 - 10 | 1784 | 59 | |
| XM_022425269.1 | XP_022280977.1 | 9 - 10 | 1928 | 59 | ||
| XM_022425270.1 | XP_022280978.1 | 9 - 10 | 1928 | 59 | ||
| XM_022425271.1 | XP_022280979.1 | 9 - 10 | 1928 | 59 | ||
| XM_022425272.1 | XP_022280980.1 | 8 - 9 | 1754 | 59 | ||
| GenBank mRNA | AB923814.1 | - | 9 - 10 | 1592 | 59 | |
More Information
Set Membership: |
|

SLC22A5
Genomic Map