Assay Details
| Gene Symbol: SPARCL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005639161.3 | XP_005639218.1 | 6 - 7 | 1834 | 71 | |
| XM_005639162.3 | XP_005639219.1 | 7 - 8 | 1961 | 71 | ||
| XM_014109990.2 | XP_013965465.1 | 7 - 8 | 1956 | 71 | ||
| XM_014109991.2 | XP_013965466.1 | 5 - 6 | 1660 | 71 | ||
| XM_022413699.1 | XP_022269407.1 | 6 - 7 | 1788 | 71 | ||
| XM_022413700.1 | XP_022269408.1 | 6 - 7 | 1783 | 71 | ||
More Information
Set Membership: |
|

SPARCL1
Genomic Map