Assay Details
| Gene Symbol: IDH3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005618436.3 | XP_005618493.1 | 10 - 11 | 1030 | 69 | |
| XM_022417022.1 | XP_022272730.1 | 9 - 10 | 995 | 69 | ||
| XM_536213.6 | XP_536213.2 | 10 - 11 | 1093 | 69 | ||
More Information
Set Membership: |
|

IDH3A
Genomic Map