Assay Details
| Gene Symbol: CERT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022416787.1 | XP_022272495.1 | 1578 | 138 | ||
| XM_022416788.1 | XP_022272496.1 | 1524 | 138 | |||
| XM_022416789.1 | XP_022272497.1 | 1452 | 138 | |||
| XM_022416790.1 | XP_022272498.1 | 1446 | 138 | |||
More Information
Set Membership: |
|

CERT1
Genomic Map