Assay Details
| Gene Symbol: MAP3K20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022414748.1 | XP_022270456.1 | 3 - 4 | 378 | 92 | |
| XM_022414749.1 | XP_022270457.1 | 3 - 4 | 486 | 92 | ||
| XM_022414750.1 | XP_022270458.1 | 3 - 4 | 378 | 92 | ||
| XM_535966.6 | XP_535966.2 | 4 - 5 | 629 | 92 | ||
More Information
Set Membership: |
|

MAP3K20
Genomic Map