Assay Details
| Gene Symbol: GTF2H2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005617466.3 | XP_005617523.1 | 8 - 9 | 795 | 115 | |
| XM_022410307.1 | XP_022266015.1 | 7 - 8 | 441 | 115 | ||
| XM_535266.6 | XP_535266.2 | 8 - 9 | 727 | 115 | ||
More Information
Set Membership: |
|

GTF2H2
Genomic Map