Assay Details
| Gene Symbol: SLC25A38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005634266.3 | XP_005634323.1 | 2 - 3 | 404 | 66 | |
| XM_022408497.1 | XP_022264205.1 | 2 - 3 | 223 | 66 | ||
| XM_855226.5 | XP_860319.1 | 2 - 3 | 419 | 66 | ||
More Information
Set Membership: |
|

SLC25A38
Genomic Map