Assay Details
| Gene Symbol: C2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014118342.2 | XP_013973817.2 | 13 - 14 | 1848 | 82 | |
| XM_022425858.1 | XP_022281566.1 | 14 - 15 | 1987 | 82 | ||
| XM_022425859.1 | XP_022281567.1 | 13 - 14 | 1867 | 82 | ||
More Information
Set Membership: |
|

C2
Genomic Map