Assay Details
| Gene Symbol: BTF3L4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003639515.4 | XP_003639563.1 | 5 - 6 | 573 | 121 | |
| XM_005629004.3 | XP_005629061.1 | 5 - 6 | 575 | 121 | ||
| XM_014119448.2 | XP_013974923.1 | 4 - 5 | 445 | 121 | ||
| XM_014119449.2 | XP_013974924.1 | 4 - 5 | 494 | 121 | ||
| XM_022427914.1 | XP_022283622.1 | 5 - 6 | 562 | 121 | ||
| XM_022427915.1 | XP_022283623.1 | 5 - 6 | 465 | 121 | ||
| XM_022427916.1 | XP_022283624.1 | 4 - 5 | 459 | 121 | ||
More Information
Set Membership: |
|

BTF3L4
Genomic Map