Assay Details
| Gene Symbol: BUD31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005621082.3 | XP_005621139.1 | 667 | 80 | ||
| XM_005621084.3 | XP_005621141.1 | 590 | 80 | |||
| XM_022419713.1 | XP_022275421.1 | 672 | 80 | |||
| XM_022419714.1 | XP_022275422.1 | 455 | 80 | |||
| XM_855762.5 | XP_860855.1 | 583 | 80 | |||
More Information
Set Membership: |
|

BUD31
Genomic Map