Assay Details
| Gene Symbol: HHATL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005634308.3 | XP_005634365.1 | 6 - 7 | 1222 | 79 | |
| XM_005634309.3 | XP_005634366.1 | 6 - 7 | 839 | 79 | ||
| XM_014106543.2 | XP_013962018.1 | 6 - 7 | 847 | 79 | ||
| XM_022408525.1 | XP_022264233.1 | 5 - 6 | 2233 | 79 | ||
| XM_022408526.1 | XP_022264234.1 | 4 - 5 | 2121 | 79 | ||
| XM_022408527.1 | XP_022264235.1 | 4 - 5 | 593 | 79 | ||
| XM_022408528.1 | XP_022264236.1 | 5 - 6 | 725 | 79 | ||
| XM_022408529.1 | XP_022264237.1 | 4 - 5 | 958 | 79 | ||
| XM_022408530.1 | XP_022264238.1 | 4 - 5 | 611 | 79 | ||
| XM_534234.5 | XP_534234.2 | 6 - 7 | 904 | 79 | ||
More Information
Set Membership: |
|

HHATL
Genomic Map