Assay Details
| Gene Symbol: RNF34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022410690.1 | XP_022266398.1 | 332 | 90 | ||
| XM_022410691.1 | XP_022266399.1 | 242 | 90 | |||
| XM_022410692.1 | XP_022266400.1 | 506 | 90 | |||
| XM_022410693.1 | XP_022266401.1 | 325 | 90 | |||
| XM_022410694.1 | XP_022266402.1 | 304 | 90 | |||
| XM_534667.6 | XP_534667.3 | 374 | 90 | |||
More Information
Set Membership: |
|

RNF34
Genomic Map