Assay Details
| Gene Symbol: FOSL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005630291.3 | XP_005630348.1 | 3 - 4 | 673 | 84 | |
| XM_005630292.3 | XP_005630349.1 | 3 - 4 | 833 | 84 | ||
| XM_022404596.1 | XP_022260304.1 | 2 - 3 | 1982 | 84 | ||
| XM_845624.5 | XP_850717.1 | 2 - 3 | 1207 | 84 | ||
| XM_857127.5 | XP_862220.1 | 2 - 3 | 967 | 84 | ||
More Information
Set Membership: |
|

FOSL2
Genomic Map