Assay Details
| Gene Symbol: SAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014111920.2 | XP_013967395.1 | 2 - 3 | 1493 | 88 | |
| XM_537982.6 | XP_537982.2 | 3 - 4 | 565 | 88 | ||
More Information
Set Membership: |
|

SAT1
Genomic Map