Assay Details
| Gene Symbol: GALK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005638397.3 | XP_005638454.1 | 4 - 5 | 519 | 77 | |
| XM_005638398.2 | XP_005638455.1 | 4 - 5 | 519 | 77 | ||
| XM_005638399.2 | XP_005638456.1 | 4 - 5 | 520 | 77 | ||
| XM_022412980.1 | XP_022268688.1 | 4 - 5 | 520 | 77 | ||
| XM_544673.5 | XP_544673.2 | 4 - 5 | 518 | 77 | ||
More Information
Set Membership: |
|

GALK2
Genomic Map