Assay Details
| Gene Symbol: POLL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014108883.2 | XP_013964358.1 | 5 - 6 | 1782 | 66 | |
| XM_014108884.1 | XP_013964359.1 | 2 - 3 | 239 | 66 | ||
| XM_022411671.1 | XP_022267379.1 | 6 - 7 | 1632 | 66 | ||
| XM_022411672.1 | XP_022267380.1 | 5 - 6 | 1782 | 66 | ||
| XM_845558.4 | XP_850651.1 | 6 - 7 | 1220 | 66 | ||
More Information
Set Membership: |
|

POLL
Genomic Map