Assay Details
| Gene Symbol: MPIG6B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022425850.1 | XP_022281558.1 | 3 - 4 | 904 | 74 | |
| XM_844693.5 | XP_849786.2 | 4 - 5 | 972 | 74 | ||
More Information
Set Membership: |
|

MPIG6B
Genomic Map