Assay Details
| Gene Symbol: CRYGS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080901.2 | NP_001074370.1 | 2 - 3 | 296 | 125 | |
| GenBank mRNA | EF191196.1 | - | 2 - 3 | 298 | 125 | |
More Information
Set Membership: |
|

CRYGS
Genomic Map