Assay Details
| Gene Symbol: LCN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022423769.1 | XP_022279477.1 | 3 - 4 | 326 | 93 | |
| XM_022423770.1 | XP_022279478.1 | 3 - 4 | 418 | 93 | ||
More Information
Set Membership: |
|

LCN2
Genomic Map