Assay Details
| Gene Symbol: FDPS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005622722.3 | XP_005622779.1 | 9 - 10 | 1156 | 148 | |
| XM_005622727.3 | XP_005622784.1 | 7 - 8 | 800 | 148 | ||
| XM_005622731.3 | XP_005622788.1 | 6 - 7 | 620 | 148 | ||
| XM_014115340.2 | XP_013970815.1 | 7 - 8 | 1236 | 148 | ||
| XM_014115341.2 | XP_013970816.1 | 7 - 8 | 1216 | 148 | ||
| XM_014115342.2 | XP_013970817.1 | 9 - 10 | 1191 | 148 | ||
| XM_014115344.2 | XP_013970819.1 | 7 - 8 | 818 | 148 | ||
| XM_022421289.1 | XP_022276997.1 | 6 - 7 | 1157 | 148 | ||
| XM_022421290.1 | XP_022276998.1 | 8 - 9 | 973 | 148 | ||
| XM_022421291.1 | XP_022276999.1 | 8 - 9 | 952 | 148 | ||
More Information
Set Membership: |
|

FDPS
Genomic Map