Assay Details
| Gene Symbol: SLC22A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286961.1 | NP_001273890.1 | 1232 | 140 | ||
| GenBank mRNA | AY028623.1 | - | 444 | 140 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC22A2| Gene Symbol: SLC22A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286961.1 | NP_001273890.1 | 1232 | 140 | ||
| GenBank mRNA | AY028623.1 | - | 444 | 140 | ||
Set Membership: |
|