Assay Details
| Gene Symbol: SMG9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005616489.3 | XP_005616546.1 | 4 - 5 | 632 | 59 | |
| XM_022425190.1 | XP_022280898.1 | 4 - 5 | 633 | 59 | ||
| XM_022425194.1 | XP_022280902.1 | 4 - 5 | 590 | 59 | ||
| XM_022425198.1 | XP_022280906.1 | 3 - 4 | 502 | 59 | ||
| XM_022425212.1 | XP_022280920.1 | 3 - 4 | 477 | 59 | ||
| XM_022425216.1 | XP_022280924.1 | 2 - 3 | 345 | 59 | ||
| XM_533650.6 | XP_533650.2 | 4 - 5 | 631 | 59 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SMG9
Genomic Map