Assay Details
| Gene Symbol: GALNT12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022425645.1 | XP_022281353.1 | 6 - 7 | 1026 | 120 | |
| XM_532008.7 | XP_532008.3 | 6 - 7 | 1268 | 120 | ||
More Information
Set Membership: |
|

GALNT12
Genomic Map