Assay Details
| Gene Symbol: SPAG5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005624855.3 | XP_005624912.1 | 20 - 21 | 3311 | 121 | |
| XM_537743.7 | XP_537743.4 | 20 - 21 | 3313 | 121 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SPAG5
Genomic Map