Assay Details
| Gene Symbol: FRMD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005623395.3 | XP_005623452.1 | 9 - 10 | 1117 | 70 | |
| XM_022422002.1 | XP_022277710.1 | 11 - 12 | 1399 | 70 | ||
| XM_022422003.1 | XP_022277711.1 | 9 - 10 | 1083 | 70 | ||
| XM_022422004.1 | XP_022277712.1 | 9 - 10 | 1080 | 70 | ||
| XM_858957.5 | XP_864050.1 | 9 - 10 | 1093 | 70 | ||
More Information
Set Membership: |
|

FRMD6
Genomic Map