Assay Details
| Gene Symbol: HIPK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005616559.3 | XP_005616616.1 | 1 - 2 | 641 | 139 | |
| XM_533668.5 | XP_533668.2 | 1 - 2 | 641 | 139 | ||
More Information
Set Membership: |
|

HIPK4
Genomic Map