Assay Details
| Gene Symbol: C1H9orf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014117227.2 | XP_013972702.1 | 6 | 1725 | 169 | |
| XM_022420369.1 | XP_022276077.1 | 6 | 1725 | 169 | ||
| XM_022420373.1 | XP_022276081.1 | 6 | 1725 | 169 | ||
| XM_022420383.1 | XP_022276091.1 | 6 | 1725 | 169 | ||
More Information
Set Membership: |
|

C1H9orf3
Genomic Map