Assay Details
| Gene Symbol: CEP19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014110300.2 | XP_013965775.1 | 3 | 565 | 101 | |
| XM_014110303.2 | XP_013965778.1 | 3 | 561 | 101 | ||
| XM_846861.5 | XP_851954.1 | 3 | 665 | 101 | ||
More Information
Set Membership: |
|

CEP19
Genomic Map