Assay Details
| Gene Symbol: GBGT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003193.1 | NP_001003193.1 | 6 | 798 | 171 | |
| XM_014116857.2 | XP_013972332.1 | 5 | 943 | 171 | ||
| GenBank mRNA | U66140.1 | - | 6 | 798 | 171 | |
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

GBGT1
Genomic Map