Assay Details
| Gene Symbol: TOR2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022423790.1 | XP_022279498.1 | 4 | 608 | 111 | |
| XM_022423791.1 | XP_022279499.1 | 4 | 944 | 111 | ||
| XM_846407.6 | XP_851500.3 | 5 | 1063 | 111 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TOR2A
Genomic Map