Assay Details
| Gene Symbol: OMA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022418799.1 | XP_022274507.1 | 4 - 5 | 669 | 123 | |
| XM_546689.6 | XP_546689.2 | 5 - 6 | 1136 | 123 | ||
More Information
Set Membership: |
|

OMA1
Genomic Map