Assay Details
| Gene Symbol: WASF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005627707.3 | XP_005627764.1 | 8 - 9 | 1659 | 84 | |
| XM_005627708.3 | XP_005627765.1 | 8 - 9 | 1687 | 84 | ||
| XM_014118199.2 | XP_013973674.1 | 8 - 9 | 1684 | 84 | ||
| XM_014118200.2 | XP_013973675.1 | 8 - 9 | 1681 | 84 | ||
| XM_022426441.1 | XP_022282149.1 | 7 - 8 | 1883 | 84 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

WASF1
Genomic Map