Assay Details
| Gene Symbol: TAF5L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022417426.1 | XP_022273134.1 | 459 | 83 | ||
| XM_546100.3 | XP_546100.2 | 280 | 83 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: TAF5L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022417426.1 | XP_022273134.1 | 459 | 83 | ||
| XM_546100.3 | XP_546100.2 | 280 | 83 | |||
Set Membership: |
|