Assay Details
| Gene Symbol: PIP5K1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005615884.3 | XP_005615941.1 | 6 - 7 | 668 | 160 | |
| XM_005615885.3 | XP_005615942.1 | 3 - 4 | 411 | 160 | ||
| XM_014117668.2 | XP_013973143.1 | 6 - 7 | 667 | 160 | ||
| XM_014117673.2 | XP_013973148.1 | 3 - 4 | 330 | 160 | ||
| XM_022421435.1 | XP_022277143.1 | 5 - 6 | 481 | 160 | ||
| XM_022421438.1 | XP_022277146.1 | 5 - 6 | 512 | 160 | ||
More Information
Set Membership: |
|

PIP5K1B
Genomic Map