Assay Details
Gene Symbol: ERGIC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022417688.1 | XP_022273396.1 | 12 - 13 | 2599 | 151 | |
XM_022417689.1 | XP_022273397.1 | 10 - 11 | 944 | 151 | ||
XM_022417690.1 | XP_022273398.1 | 10 - 11 | 932 | 151 | ||
XM_022417691.1 | XP_022273399.1 | 10 - 11 | 962 | 151 | ||
XM_847798.3 | XP_852891.1 | 9 - 10 | 827 | 151 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
