Assay Details
| Gene Symbol: LCMT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005621396.2 | XP_005621453.1 | 8 - 9 | 925 | 147 | |
| XM_005621397.2 | XP_005621454.1 | 6 - 7 | 760 | 147 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

LCMT1
Genomic Map