Assay Details
| Gene Symbol: COX18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005628280.3 | XP_005628337.2 | 1 - 2 | 350 | 150 | |
| XM_539312.5 | XP_539312.4 | 1 - 2 | 351 | 150 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

COX18
Genomic Map