Assay Details
| Gene Symbol: DAZL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005634380.2 | XP_005634437.1 | 2 - 3 | 278 | 144 | |
| XM_022408600.1 | XP_022264308.1 | 2 - 3 | 371 | 144 | ||
| XM_534251.5 | XP_534251.3 | 2 - 3 | 582 | 144 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005634380.2 transcripts.
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

DAZL
Genomic Map