Assay Details
| Gene Symbol: S1PR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005621856.3 | XP_005621913.1 | 2 | 1155 | 82 | |
| XM_005621857.3 | XP_005621914.1 | 2 | 1302 | 82 | ||
| XM_005621858.3 | XP_005621915.1 | 2 | 1071 | 82 | ||
| XM_547259.6 | XP_547259.2 | 2 | 1301 | 82 | ||
More Information
Set Membership: |
|

S1PR1
Genomic Map