Assay Details
| Gene Symbol: GNAL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005623091.3 | XP_005623148.2 | 2 - 3 | 241 | 138 | |
| XM_014115587.2 | XP_013971062.1 | 3 - 4 | 333 | 138 | ||
| XM_014115588.2 | XP_013971063.1 | 4 - 5 | 973 | 138 | ||
| XM_022421638.1 | XP_022277346.1 | 5 - 6 | 1183 | 138 | ||
| XM_022421639.1 | XP_022277347.1 | 5 - 6 | 873 | 138 | ||
| XM_022421640.1 | XP_022277348.1 | 4 - 5 | 626 | 138 | ||
| XM_022421641.1 | XP_022277349.1 | 4 - 5 | 936 | 138 | ||
More Information
Set Membership: |
|

GNAL
Genomic Map