Assay Details
| Gene Symbol: GNB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003004.1 | NP_001003004.1 | 3 - 4 | 465 | 144 | |
| XM_005636669.3 | XP_005636726.1 | 3 - 4 | 144 | |||
| GenBank mRNA | DQ672621.1 | - | 3 - 4 | 466 | 144 | |
| EU285563.1 | - | 3 - 4 | 466 | 144 | ||
| U52916.1 | - | 3 - 4 | 465 | 144 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

GNB3
Genomic Map