Assay Details
| Gene Symbol: H3-3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_537232.4 | XP_537232.1 | 1 - 2 | 83 | 60 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_537232.4 transcripts.
Set Membership: |
|

H3-3A
Genomic Map