Assay Details
| Gene Symbol: NR4A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003227.1 | NP_001003227.1 | 2 - 3 | 944 | 152 | |
| XM_005636676.3 | XP_005636733.1 | 4 - 5 | 1209 | 152 | ||
| XM_014108245.2 | XP_013963720.1 | 2 - 3 | 1135 | 152 | ||
| XM_014108246.2 | XP_013963721.1 | 3 - 4 | 1111 | 152 | ||
| XM_022410843.1 | XP_022266551.1 | 3 - 4 | 1137 | 152 | ||
| GenBank mRNA | X97226.1 | - | 944 | 152 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

NR4A1
Genomic Map