Assay Details
| Gene Symbol: IARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014118130.2 | XP_013973605.1 | 3 - 4 | 310 | 102 | |
| XM_022422371.1 | XP_022278079.1 | 3 - 4 | 419 | 102 | ||
| XM_541332.6 | XP_541332.3 | 3 - 4 | 444 | 102 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

IARS1
Genomic Map