Assay Details
| Gene Symbol: PAK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022414036.1 | XP_022269744.1 | 10 - 11 | 933 | 87 | |
| XM_844339.6 | XP_849432.1 | 11 - 12 | 1200 | 87 | ||
More Information
Set Membership: |
|

PAK2
Genomic Map