Assay Details
| Gene Symbol: PRKAG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005636885.3 | XP_005636942.3 | 11 - 12 | 1067 | 110 | |
| XM_014108393.2 | XP_013963868.2 | 11 - 12 | 1142 | 110 | ||
| XM_014108394.2 | XP_013963869.1 | 11 - 12 | 1084 | 110 | ||
| XM_022410935.1 | XP_022266643.1 | 11 - 12 | 1130 | 110 | ||
| XM_022410936.1 | XP_022266644.1 | 11 - 12 | 1028 | 110 | ||
| XM_022410937.1 | XP_022266645.1 | 11 - 12 | 1014 | 110 | ||
| XM_022410938.1 | XP_022266646.1 | 10 - 11 | 990 | 110 | ||
| XM_543685.6 | XP_543685.2 | 11 - 12 | 1009 | 110 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PRKAG1
Genomic Map