Assay Details
| Gene Symbol: MAPK6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005638418.3 | XP_005638475.1 | 3 - 4 | 1554 | 104 | |
| XM_005638419.3 | XP_005638476.1 | 4 - 5 | 1626 | 104 | ||
| XM_858073.5 | XP_863166.2 | 3 - 4 | 1486 | 104 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

MAPK6
Genomic Map